EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
The construction of high-nuclearity isopolyoxoniobates with pentagonal building blocks: [HNb27O76]16- and [H10Nb31O93(CO3)]23-.
PMID 19946917