EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
An enantiocomplementary dirigent protein for the enantioselective laccase-catalyzed oxidative coupling of phenols.
PMID 19946920
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06709
Pinoresinol, analytical standard
C20H22O6
40674
Pinoresinol, ≥95.0% (HPLC)
C20H22O6