EMAIL THIS PAGE TO A FRIEND
Laboratory investigation; a journal of technical methods and pathology
Runx2 regulates survivin expression in prostate cancer cells.
PMID 19949374
Abstract
Previously we described that bone morphogenetic protein-7 (BMP7) could protect prostate cancer C4-2B cells from serum starvation-induced apoptosis via survivin induction. Here, for the first time, we identify Runx2 as a key regulator of survivin transcription. In C4-2B cells grown normally, suppression of Runx2 reduced survivin expression. Using ChIP assays, two regions of the survivin promoter, -1953 to -1812 (I) and -1485 to -1119 (II) encompassing consensus Runx-binding sites were examined. Runx2 was found to be associated with both regions, with a stronger affinity to region-I. In serum-starved cells neither region was occupied, but BMP7 restored association to region-II and not region-I. In reporter assays, transcription activity by BMP7 was significantly reduced when sequences including binding sites of region-II were deleted. Additionally, Runx2 expression was enhanced by BMP7 in these cells. Along with a strong survivin expression, a trend in increased Runx2 expression in human prostate cancer cells and tissues was noted. In the conditional Pten-knockout mouse, Runx2 level increased with growth of prostate tumor. The data define a novel role of Runx2 in regulating survivin expression in malignant epithelial cells and identify it as a critical factor in BMP signaling that protects cancer cells against apoptosis.