EMAIL THIS PAGE TO A FRIEND
Psychiatry research
Gender, personality, and serotonin-2A receptor binding in healthy subjects.
PMID 19959344
Abstract
The vulnerability to mood disorders, impulsive-aggression, eating disorders, and suicidal behavior varies greatly with gender, and may reflect gender differences in central serotonergic function. We investigated the relationships of gender, mood, impulsivity, aggression and temperament to 5HT(2A) receptor binding in 21 healthy subjects using [18F]altanserin and PET neuroimaging. Binding potentials in pre-defined regions-of-interest (ROI) were calculated using the Logan graphical method, corrected for partial volume effects, and compared by gender with age co-varied. SPM analysis was used for voxel level comparisons. Altanserin binding (BP(P)) was greater in male than female subjects in the following nine ROIs: hippocampus (HIP) and Lt. HIP, lateral orbital frontal cortex (LOF) and Lt. LOF, left medial frontal cortex (Lt. MFC), left medial temporal cortex (Lt. MTC), left occipital cortex (Lt. OCC), thalamus (THL) and Lt. THL. Differences in Lt. HIP and Lt. MTL remained significant after Bonferroni correction. Gender differences were noted in the co-variation of psychological traits with BP(P) values in specific ROIs. Among males alone, aggression was negatively correlated with BP(P) values in Lt. LOF and Lt. MFC, and Suspiciousness positively correlated in LOF, Lt. LOF and Lt. MFC. Among female subjects alone, Negativism was positively correlated with BP(P) values in HIP, and Verbal Hostility in Lt. HIP. Altanserin binding in Lt. MTC was positively correlated with Persistence, with no significant gender effect. Gender differences in 5HT(2A) receptor function in specific ROIs may mediate expression of psychological characteristics such as aggression, suspiciousness and negativism. Future studies of 5HT(2A) receptor function and its relationship to behavior should control for gender.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8106
Altanserin hydrochloride hydrate, ≥98% (HPLC), solid
C22H22FN3O2S · HCl · xH2O