EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Studies on chemical constituents of fructus broussonetiae].
PMID 19960944
Abstract
To study the chemical constituents of Fructus Broussonetiae. Column chromatography with silic gel was employed to isolate and purify the 80% alcohol extract of Fructus Broussonetia, and the constituents were identified by spectral methods. Six compounds were isolated from 80% ethanol extract. Their structures were identified as Isoterihanine (1), Chelerythrine (2), Trillin (3), Sucrose (4), beta-sitosterol (5) and Fucosterol (6). These compounds are isolated from Fructus Broussonetiae for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5379
Fucosterol, ≥93%
C29H48O