EMAIL THIS PAGE TO A FRIEND
Natural product communications
Cytotoxic metabolites produced by Alternaria no.28, an endophytic fungus isolated from Ginkgo biloba.
PMID 19967976
Abstract
From the medicinal plant Ginkgo biloba the fungal endophyte Alternaria no.28 was isolated. Extract of the fungus grown in liquid culture media exhibited marked cytotoxic activity when tested in vitro against brine shrimp (Artemia salina). Eight compounds were isolated from the extract of cultures of this endophytic fungus and were elucidated as alterperylenol (1), altertoxin I (2), alternariol (3), alternariol monomethyl ether (4), tenuazonic acid (5) and its derivative (6), together with ergosterol and ergosta-4, 6, 8, 22-tetraen-3-one by means of spectroscopic analysis. Among them, both 5 and 6 showed significant cytotoxic effects in the brine shrimp bioassy, with mortality rates of 73.6% and 68.9%, respectively, at a concentration of 10 microg x mL(-1), and they were first isolated from endophytic fungi.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
37018
Tenuazonic acid, analytical standard
C10H15NO3
T3408
Tenuazonic acid copper salt from Alternaria alternata
C10H14NO3 · 0.5Cu