EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Phosphate release contributes to the rate-limiting step for unwinding by an RNA helicase.
PMID 19969541
Abstract
RNA helicases function in numerous aspects of RNA biology. These enzymes are RNA-stimulated ATPases that translocate on RNA and unwind or remodel structured RNA in an ATP-dependent fashion. How ATP and the ATPase cycle fuel the work performed by helicases is not completely clear. The hepatitis C virus RNA helicase, NS3, is an important model system for this class of enzymes. NS3 binding to a single-/double-strand RNA or DNA junction leads to ATP-independent melting of the duplex and formation of a complex capable of ATP-dependent unwinding by using a spring-loaded mechanism. We have established an RNA substrate for NS3 that can be unwound in a single sub-step. Our studies are consistent with a model in which a single ATP binding and/or hydrolysis event sets the unwinding spring and phosphate dissociation contributes to release of the spring, thereby driving the power stroke used for unwinding.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G7627
Guanosine 5′-monophosphomorpholidate 4-morpholine-N,N′-dicyclohexylcarboxamidine salt, ~95%
C14H21N6O8P · C17H31N3O