EMAIL THIS PAGE TO A FRIEND
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections
O-aminoazotoluene suppresses hepatocyte proliferation in inbred mice susceptible to hepatocarcinogenesis.
PMID 19994796
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
121568
Fast Garnet GBC base, 97%
C14H15N3
31629
o-Aminoazotoluene, analytical standard
C14H15N3