EMAIL THIS PAGE TO A FRIEND
Drug and chemical toxicology
A 90-day intravenous administration toxicity study of CaO-SiO2-P2O5-B2O3 glass-ceramics (BGS-7) in rat.
PMID 19995308
Abstract
CaO-SiO(2)-P(2)O(5)-B(2)O(3) glass ceramics directly bond to bone and have potential use as a bone substitute material. The present study evaluated the toxicity from subchronic intravenous administration of CaO-SiO(2)-P(2)O(5)-B(2)O(3) glass ceramics to male and female Sprague-Dawley rats. The rats were divided into four groups, each consisting of 10 male and 10 female rats, and administered different amounts of CaO-SiO(2)-P(2)O(5)-B(2)O(3) glass ceramics (aqueous extract 5, 2.5, and 1.25 mL/kg body weight/day and saline) 7 days per week for 90 consecutive days. During the experiment, no deaths were observed in any groups, and there were no remarkable changes in clinical signs, body weight, food and water intake, hematological and serum biochemical parameters, organ weight, and histopathological findings between the control and treated groups. The results show no adverse toxic effects of CaO-SiO(2)-P(2)O(5)-B(2)O(3) glass ceramics (aqueous extract 5 mL/kg body weight/day) to rats of either sex.