EMAIL THIS PAGE TO A FRIEND
Laeknabladid
[Allergy and skin infection after use of temporary henna tattoo--case report].
PMID 19996466
Abstract
We describe a case of a 10 year old boy who developed an contact dermatitis to black henna tattoo. Sixteen days later he was brought to the emergency department because of an presumed superinfection by S. aureus. The infection was successfully treated with dicloxacillin and the allergic reaction with bethametasone ointment and tablets. The use of pure henna is legal but has been proven to be harmful in animal experiments. Para-phenylendiamine (PPD) has been mixed with henna to achieve a darker colour and to decrease the treatment time and is well known to cause allergic reactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9016
Dicloxacillin sodium salt monohydrate
C19H16Cl2N3NaO5S·H2O