EMAIL THIS PAGE TO A FRIEND
Southern medical journal
Drug-induced liver injury caused by the histrelin (Vantus) subcutaneous implant.
PMID 19996852
Abstract
Drug-induced liver injury (DILI) is the leading cause of acute hepatic failure in the United States. Up to 13% of acute liver failure cases occur due to drugs other than acetaminophen. This clinical diagnosis, made after other causes of liver injury have been excluded, requires establishing a causal relationship between drug exposure and liver injury. The case of a patient with liver injury following a subcutaneous histrelin (Vantus) implant as therapy for advanced prostate cancer is presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L2761
[des-Gly10, D-His(Bzl)6]-LH-RH ethylamide, ≥97% (HPLC), powder
C66H86N18O12