EMAIL THIS PAGE TO A FRIEND
Optics express
Simultaneous reference and differential waveform acquisition in time-resolved terahertz spectroscopy.
PMID 19997441
Abstract
We present a new method for data acquisition in time-resolved terahertz spectroscopy experiments. Our approach is based on simultaneous collection of reference and differential THz scans. Both the optical THz generation beam and the pump beam are modulated at two different frequencies that are not harmonic with respect to each other. Our method allows not only twice as fast data acquisition but also minimization of noise connected to slowly varying laser power fluctuations and timing instabilities. Our use of the nonlinear crystal N-benzyl-2-methyl-4-nitroaniline (BNA) enables time-resolved THz spectroscopy to beyond 5 THz, thereby highlighting that the presented method is especially valuable at higher frequencies where phase errors in the data acquisition become increasingly important.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
185310
4-Nitroaniline, ≥99%
C6H6N2O2
31569
4-Nitroaniline, analytical standard
C6H6N2O2