EMAIL THIS PAGE TO A FRIEND
Journal of environmental sciences (China)
Isolation and characterization of Sporobolomyces sp. LF1 capable of degrading chlorimuron-ethyl.
PMID 19999974
Abstract
A yeast strain which was capable of degrading sulfonylurea herbicide chlorimuron-ethyl named as LF1 was isolated from a chlorimuron-ethyl contaminated soil near the warehouse of the factory producing chlorimuron-ethyl in Shenyang City, Northeast China. The strain was identified as Sporobolomyces sp., based on its morphological and physiological characteristics and the phylogenetic analysis of 18S rRNA gene sequence. So far, this is the only yeast strain of Sporobolomyces sp. which is able to degrade chlorimuron-ethyl. Incubation tests showed that when the initial concentration of chlorimuron-ethyl in culture was 5 mg/L, LFI could degrade more than 77% of the herbicide after incubation for 4 d at 30 degrees C. The possible mechanism of chlorimuron-ethyl degradation by LF1 could be the acidic hydrolysis caused by the acids from the metabolism of the yeast strain. Further study should be conducted to examine the pathways of chlorimuron-ethyl degradation by LF1 and to approach the feasibility of using LF1 to degrade the chlorimuron-ethyl in soil system.