EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Enzymatic production of (-)-indolactam V by LtxB, a cytochrome P450 monooxygenase.
PMID 20000453
Abstract
The P450 cytochrome monooxygenase gene, ltxB, was cloned and overexpressed in Escherichia coli as a 6xHis-tagged protein. The resulting recombinant LtxB was purified by Ni-NTA affinity chromatography and characterized biochemically. Purified LtxB demonstrated typical cytochrome P450 spectroscopic properties including substrate-induced transition from a low-spin (lambdamax=414 nm) to high-spin state (lambdamax=386 nm) upon incubation with N-methyl-L-valyl-L-tryptophanol. The catalytic activity of LtxB was verified by demonstrating the oxidation/cyclization of N-methyl-L-valyl-L-tryptophanol to (-)-indolactam V. LtxB shows a relaxed specificity for analogue substrates in which the valyl group is substituted for other aliphatic groups. The relaxed substrate specificity of LtxB, along with the relaxed specificity of the prenyltransferase, LtxC, allowed for the enzymatic production of a series of (-)-indolactam V and lyngbyatoxin analogues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I0661
(−)-Indolactam V, ≥96% (HPLC)
C17H23N3O2