EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Quantification of (15)N-nitrate in urine with gas chromatography combustion isotope ratio mass spectrometry to estimate endogenous NO production.
PMID 20000695
Abstract
The use of stable isotope labeled substrates and subsequent analysis of urinary nitrate, forms a noninvasive test for evaluation of the in vivo NO metabolism. The present paper describes a new method for simultaneous quantification of (15)N-nitrate and total nitrate with gas chromatography combustion isotope ratio mass spectrometry (GC-C-IRMS). Nitrate, isolated from urine with a nitrate selective resin, was reduced to nitrite using copperized cadmium. Subsequently, Sudan I was formed by diazotation. Sudan II was added as internal standard, and both molecules were analyzed with GC-C-IRMS as tert-butyldimethylsilyl derivatives. The accuracy was determined during a recovery study of two different known nitrate concentrations and two (15)N-enrichments. A recovery of 101.6% and 103.9% for total nitrate and 107.6% and 91.2% for (15)N-nitrate was obtained, respectively. The validated method was applied on complete 72 h urine collections after intravenous administration of (15)N-nitrate and (15)N-arginine in humans. On average, 51.8% (47.0-71.0%) of administered (15)N-nitrate was excreted, while 0.68% (0.44-1.17%) of (15)N-arginine was metabolized to nitrate. In conclusion, this method can be used for accurate simultaneous determination of (15)N-nitrate and total nitrate concentrations in urine and can be applied in clinical studies for noninvasive evaluation of NO metabolism in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
199656
Sudan II, Dye content 90 %
C18H16N2O
07937
Sudan II, analytical standard
C18H16N2O