EMAIL THIS PAGE TO A FRIEND
Biotechnic & histochemistry : official publication of the Biological Stain Commission
De-staining and re-staining mucins in formalin fixed paraffin sections.
PMID 20001228
Abstract
Re-staining of formalin fixed paraffin sections sometimes is required and this requires prior de-staining. Some simple and effective protocols for de-staining are described. Mucihematoxylin and mucicarmine can be removed with acid alcohol. Zirconyl hematoxylin can be removed with periodic acid or Sinha's fixative. Alcian blue can be removed with 5% trifluoroacetic acid in dichloromethane. Colloidal iron can be bleached in 1% household bleach in alcohol. PAS can be removed with hydrogen peroxide or ammonium hydroxide. With few exceptions, de-stained sections can be re-stained with mucihematoxylin, PAS or Gabe's trichrome.