EMAIL THIS PAGE TO A FRIEND
Journal of neurochemistry
The neuroprotective mechanism of 1-(R)-aminoindan, the major metabolite of the anti-parkinsonian drug rasagiline.
PMID 20002521
Abstract
The anti-parkinsonian drug, rasagiline [N-propargyl-1-(R)-aminoindan; Azilect(R)], is a secondary cyclic benzylamine and indane derivative, which provides irreversible, potent monoamine oxidase-B (MAO-B) inhibition and possesses neuroprotective and neurorestorative activities. A prospective clinical trial has shown that rasagiline confers significant symptomatic improvement and demonstrated alterations in Parkinson's disease progression. Rasagiline is primarily metabolized by hepatic cytochrome P-450 to form its major metabolite, 1-(R)-aminoindan, a non-amphetamine, weak reversible MAO-B inhibitor compound. Recent studies indicated the potential neuroprotective effect of 1-(R)-aminoindan, suggesting that it may contribute to the overall neuroprotective and antiapoptotic effects of its parent compound, rasagiline. This review article briefly highlights the molecular mechanisms underlying the neuroprotective properties of the active metabolite of rasagiline, 1-(R)-aminoindan, supporting the valuable potential of rasagiline for disease modification.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
479128
2-Aminoindan, 97%
C9H11N
A59522
2-Aminoindan hydrochloride, 98%
C9H11N · HCl