EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Isoxazolopyridone derivatives as allosteric metabotropic glutamate receptor 7 antagonists.
PMID 20005101
Abstract
This Letter describes the synthesis and evaluation of mGluR7 antagonists in the isoxazolopyridone series. In the course of modification in this class, novel solid support synthesis of the isoxazolopyridone scaffold was developed. Subsequent chemical modification led to the identification of several potent derivatives with improved physicochemical properties compared to a hit compound 1. Among these, 2 showed good oral bioavailability and brain penetrability, suggesting that 2 may be useful for in vivo study to elucidate the role of mGluR7.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
134198
5-Methyl-3-phenylisoxazole-4-carboxylic acid, 99%
C11H9NO3