EMAIL THIS PAGE TO A FRIEND
Journal of experimental botany
L-Glutamine inhibits beta-aminobutyric acid-induced stress resistance and priming in Arabidopsis.
PMID 20007686
Abstract
The non-protein amino acid beta-aminobutyric acid (BABA) enhances Arabidopsis resistance to microbial pathogens and abiotic stresses through potentiation of the Arabidopsis defence responses. In this study, it is shown that BABA induces the stress-induced morphogenic response (SIMR). SIMR is observed in plants exposed to sub-lethal stress conditions. Anthocyanin, a known modulator of stress signalling, was also found to accumulate in BABA-treated Arabidopsis. These data and a previous microarray study indicate that BABA induces a stress response in Arabidopsis. High concentrations of amino acids, except for L-glutamine, cause a general amino acid stress inhibition. General amino acid inhibition is prevented by the addition of L-glutamine. L-Glutamine was found to inhibit the BABA-mediated SIMR and anthocyanin accumulation, suggesting that the non-protein amino acid BABA causes a general amino acid stress inhibition in Arabidopsis. L-Glutamine also blocked BABA-induced resistance to heat stress and to the virulent bacterial pathogen Pseudomonas syringae pv. tomato DC3000. During bacterial infection, priming of the salicylic acid-dependent defence marker PR1 was abolished by L-glutamine treatment. These results indicate that L-glutamine removal of the BABA-mediated stress response is concomitant with L-glutamine inhibition of BABA priming and BABA-induced resistance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
757454
(S)-3-Aminobutyric acid, 97%
C4H9NO2
A44207
3-Aminobutanoic acid, 97%
C4H9NO2