EMAIL THIS PAGE TO A FRIEND
Circulation. Cardiovascular genetics
Autonomic function in hypertension; role of genetic variation at the catecholamine storage vesicle protein chromogranin B.
PMID 20011129