EMAIL THIS PAGE TO A FRIEND
Naunyn-Schmiedeberg's archives of pharmacology
Reversal of propoxur-induced impairment of memory and oxidative stress by 4'-chlorodiazepam in rats.
PMID 20012268
Abstract
Carbamate pesticides like propoxur have been shown to adversely affect memory and induce oxidative stress on both acute and chronic exposure. The present study was designed to explore the modulation of the effects of propoxur over cognitive function by progesterone (PROG) and 4'-chlorodiazepam (4CD). Cognitive function was assessed using step-down latency (SDL) on a passive avoidance apparatus, transfer latency (TL) on a plus maze and spatial navigation test on Morris water maze. Oxidative stress was assessed by examining brain malondialdehyde (MDA) and reduced glutathione (GSH) levels and catalase (CAT) activity. A significant reduction in SDL and prolongation of TL and spatial navigation test was found for the propoxur (10 mg/kg/d; p.o.) treated group at weeks 6 and 7 as compared with control. One-week treatment with 4CD (0.5 mg/kg/d; i.p.) antagonized the effect of propoxur on SDL, spatial navigation test as well as TL; whereas, PROG failed to modulate this effect at a dose of 15 mg/kg/d, i.p. Propoxur produced a statistically significant increase in the brain MDA levels and decrease in the brain GSH levels and CAT activity. Treatment with 4CD at the above dose attenuated the effect of propoxur on oxidative stress whereas PROG (15 mg/kg/d; i.p.) failed to influence the same. The results of the present study thus show that 4-CD has the potential to attenuate cognitive dysfunction and oxidative stress induced by toxicants like propoxur in the brain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5174
4′-Chlorodiazepam, ≥98% (TLC)
C16H12Cl2N2O