EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Preparation of frozen sections for analysis.
PMID 20012821
Abstract
The analysis of frozen tissue by antibodies can be accomplished by the quick freezing of a small tissue sample in liquid nitrogen. Super-cooled isopentane can also be used to further the preservation process. Freezing preserves the available proteins in a near-native state for their identification by antibodies raised against naturally folded proteins. The tissues are sectioned onto charged glass slides where they can be optimally fixed in weakly or non-denaturing solutions such as acetone or those that are alcohol-based. Following mild pretreatment steps to allow for antibody use with low background, (the endogenous peroxidase enzyme or oxidative compounds quenched in a hydrogen peroxide solution and available charged sites blocked by incubation in a normal serum solution) the sections are ready for antigen detection.