EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Probing the photothermal effect of gold-based nanocages with surface-enhanced Raman scattering (SERS).
PMID 20014343
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
44130
1-Dodecanethiol, purum, ≥97.0% (GC)
C12H26S