EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
DHTP ligands for the highly ortho-selective, palladium-catalyzed cross-coupling of dihaloarenes with grignard reagents: a conformational approach for catalyst improvement.
PMID 20017175