EMAIL THIS PAGE TO A FRIEND
Chemistry & biodiversity
Sinuladiterpenes A-F, new cembrane diterpenes from Sinularia flexibilis.
PMID 20020460
Abstract
Chromatographic investigation of the octocoral Sinularia flexibilis afforded six new cembrane diterpenes, sinuladiterpenes A-F (1-6, resp.), in addition to four known cembranolides, 11-episinulariolide acetate, 11-dehydrosinulariolide, 11-episinulariolide, and sinulariolide. Their structures were elucidated by spectroscopic analysis, especially 2D-NMR and HR-ESI-MS. Compound 2 exhibited significant in vitro cytotoxic activity against human colon adenocarcinoma (WiDr) cell line.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
WiDr