EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Practical asymmetric conjugate alkynylation of Meldrum's acid-derived acceptors: access to chiral beta-alkynyl acids.
PMID 20020682
Abstract
The enantioselective conjugate addition of alkynyl nucleophiles has been a long-standing challenge in synthetic chemistry. This paper describes a highly practical asymmetric conjugate alkynylation of Meldrum's acid-derived acceptors using cinchonidine (<$100/kg) as the chiral mediator. The process provides practical access to chiral beta-alkynyl acids. Noteworthy attributes of the method are its broad scope, high functional-group compatibility, and ease of scalability.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
210145
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%
C6H8O4