EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and antituberculosis activity of novel mefloquine-isoxazole carboxylic esters as prodrugs.
PMID 20022500
Abstract
5-(2,8-Bis(trifluoromethyl)quinolin-4-yloxymethyl)isoxazole-3-carboxylic acid ethyl ester (compound 3) was reported to have excellent antituberculosis activity against both replicating and non-replicating Mycobacterium tuberculosis, with a minimum inhibitory concentration (MIC) of 0.9 microM and 12.2 microM, respectively. In this study, the antituberculosis activity of compound 3 was further investigated. Its activity appeared to be very specific for organisms of the M. tuberculosis complex and it effected significant reductions of bacterial numbers in infected macrophages with an EC(90) of 4.1 microM. More importantly, the increased in vitro antituberculosis activity of the corresponding acid (compound 4) at pH 6.0 suggested that it may be active in vivo in an acidic environment produced as a consequence of inflammation in the lungs of TB patients. The fact that various ester bioisosteres of compound 3 lost anti-TB activity further suggested that the ester compound 3 may function as a prodrug. The detailed structure-activity relationships (SARs) from this study should facilitate our ultimate goal of improving the anti-TB potency of this isoxazole ester series.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
18805
6-Hydroxyindole, ≥99.0% (GC)
C8H7NO