EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Type II diacylglycerol acyltransferase from Claviceps purpurea with ricinoleic acid, a hydroxyl fatty acid of industrial importance, as preferred substrate.
PMID 20023082
Abstract
Claviceps purpurea, the fungal pathogen that causes the cereal disease ergot, produces glycerides that contain high levels of ricinoleic acid [(R)-12-hydroxyoctadec-cis-9-enoic acid] in its sclerotia. Recently, a fatty acid hydroxylase (C. purpurea FAH [CpFAH]) involved in the biosynthesis of ricinoleic acid was identified from this fungus (D. Meesapyodsuk and X. Qiu, Plant Physiol. 147:1325-1333, 2008). Here, we describe the cloning and biochemical characterization of a C. purpurea type II diacylglycerol acyltransferase (CpDGAT2) involved in the assembly of ricinoleic acid into triglycerides. The CpDGAT2 gene was cloned by degenerate RT-PCR (reverse transcription-PCR). The expression of this gene restored the in vivo synthesis of triacylglycerol (TAG) in the quadruple mutant strain Saccharomyces cerevisiae H1246, in which all four TAG biosynthesis genes (DGA1, LRO1, ARE1, and ARE2) are disrupted. In vitro enzymatic assays using microsomal preparations from the transformed yeast strain indicated that CpDGAT2 prefers ricinoleic acid as an acyl donor over linoleic acid, oleic acid, or linolenic acid, and it prefers 1,2-dioleoyl-sn-glycerol over 1,2-dipalmitoyl-sn-glycerol as an acyl acceptor. The coexpression of CpFAH with CpDGAT2 in yeast resulted in an increased accumulation of ricinoleic acid compared to the coexpression of CpFAH with the native yeast DGAT2 (S. cerevisiae DGA1 [ScDGA1]) or the expression of CpFAH alone. Northern blot analysis indicated that CpFAH is expressed solely in sclerotium cells, with no transcripts of this gene being detected in mycelium or conidial cells. CpDGAT2 was more widely expressed among the cell types examined, although expression was low in conidiospores. The high expression of CpDGAT2 and CpFAH in sclerotium cells, where high levels of ricinoleate glycerides accumulate, provided further evidence supporting the roles of CpDGAT2 and CpFAH as key enzymes for the synthesis and assembly of ricinoleic acid in C. purpurea.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R7257
Ricinoleic acid, ≥99%
C18H34O3