EMAIL THIS PAGE TO A FRIEND
Acta pharmacologica Sinica
Topical delivery of silymarin constituents via the skin route.
PMID 20023692
Abstract
Silibinin (SB), silydianin (SD), and silychristin (SC) are components of silymarin. These compounds can be used to protect the skin from oxidative stress induced by ultraviolet (UV) irradiation and treat it. To this end, the absorption of silymarin constituents via the skin was examined in the present report. Transport of SB, SD, and SC under the same thermodynamic activity through and into the skin and the effects of pH were studied in vitro using a Franz diffusion assembly. The lipophilicity increased in the order of SC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
51681
Silychristin, analytical standard
C25H22O10
30494
Silydianin, analytical standard
C25H22O10