EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Synthesis of natural products containing spiroketals via intramolecular hydrogen abstraction.
PMID 20024126
Abstract
Although known for over a quarter of a century, the oxidative radical cyclisation route to spiroketals has found limited use in natural product synthesis in comparison to classical approaches. Its successful application in this field of research forms the subject of this perspective.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1271
Narasin from Streptomyces auriofaciens, ≥98% (HPLC)
C43H72O11