EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Enantioselective iridium-catalyzed carbonyl allylation from the alcohol oxidation level via transfer hydrogenation: minimizing pre-activation for synthetic efficiency.
PMID 20024203
Abstract
Existing methods for enantioselective carbonyl allylation, crotylation and tert-prenylation require stoichiometric generation of pre-metallated nucleophiles, and often employ stoichiometric chiral modifiers. Under the conditions of transfer hydrogenation employing an ortho-cyclometallated iridium C,O-benzoate catalyst, enantioselective carbonyl allylations, crotylations and tert-prenylations are achieved in the absence of stoichiometric metallic reagents or stoichiometric chiral modifiers. Moreover, under transfer hydrogenation conditions, primary alcohols function dually as hydrogen donors and aldehyde precursors, enabling enantioselective carbonyl addition directly from the alcohol oxidation level.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
449229
Iridium, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.9% trace metals basis
Ir
357324
Iridium, foil, thickness 0.25 mm, 99.9% trace metals basis
Ir
209686
Iridium, powder, 99.9% trace metals basis
Ir
336793
Iridium, black, powder, −200 mesh, ≥99% trace metals basis
Ir
266841
Iridium, wire, diam. 0.15 mm, 99.9% trace metals basis
Ir
GF35321172
Iridium, rod, 50mm, diameter 3.0mm, as drawn, 99.9%
Ir
GF07750795
Iridium, wire reel, 0.025m, diameter 2.0mm, as drawn, 99.9%
Ir
GF44052667
Iridium, wire reel, 0.05m, diameter 0.075mm, as drawn, 99.9%
Ir
GF97286428
Iridium, wire reel, 0.05m, diameter 0.1mm, as drawn, 99.9%
Ir
GF22425375
Iridium, wire reel, 0.05m, diameter 0.8mm, as drawn, 99.9%
Ir
GF68849544
Iridium, wire reel, 0.05m, diameter 2.0mm, as drawn, 99.9%
Ir
GF06583394
Iridium, wire reel, 0.1m, diameter 0.075mm, as drawn, 99.9%
Ir
GF65585021
Iridium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.5%
Ir
GF88210447
Iridium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9%
Ir
GF26409247
Iridium, wire reel, 0.1m, diameter 0.15mm, as drawn, 99.9%
Ir
GF60803412
Iridium, wire reel, 0.1m, diameter 0.1mm, as drawn, 99.9%
Ir
GF31863607
Iridium, wire reel, 0.1m, diameter 0.25mm, as drawn, 99.9%
Ir
GF99945534
Iridium, wire reel, 0.1m, diameter 0.2mm, as drawn, 99.9%
Ir
GF36309450
Iridium, wire reel, 0.1m, diameter 0.38mm, as drawn, 99.9%
Ir
GF50407062
Iridium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9%
Ir
GF80911403
Iridium, wire reel, 0.1m, diameter 0.8mm, as drawn, 99.9%
Ir
GF22923974
Iridium, wire reel, 0.2m, diameter 0.075mm, as drawn, 99.9%
Ir
GF30370924
Iridium, wire reel, 0.2m, diameter 0.125mm, as drawn, 99.5%
Ir
GF26305130
Iridium, wire reel, 0.2m, diameter 0.125mm, as drawn, 99.9%
Ir
GF01155170
Iridium, wire reel, 0.2m, diameter 0.15mm, as drawn, 99.9%
Ir
GF00427250
Iridium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.9%
Ir
GF82950273
Iridium, wire reel, 0.2m, diameter 0.25mm, as drawn, 99.9%
Ir
GF60570176
Iridium, wire reel, 0.2m, diameter 0.2mm, as drawn, 99.9%
Ir
GF20836425
Iridium, foil, 0.025m coil, thickness 0.05mm, coil width .5mm, as rolled, 99.9%
Ir
GF10109629
Iridium, rod, 25mm, diameter 3.0mm, as drawn, 99.9%
Ir