EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Assembly of indole-2-carboxylic acid esters through a ligand-free copper-catalysed cascade process.
PMID 20024286
Abstract
A straightforward synthesis of indole-2-carboxylic esters was developed through a ligand-free copper-catalysed condensation/coupling/deformylation cascade process from 2-halo aryl aldehydes or ketones with ethyl isocyanoacetate. The reactions proceeded well for most of the 2-iodo-, bromo-, and chloro-substrates under room temperature or mild conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C88505
Cyanoacetic acid, 99%
C3H3NO2
I5109
Indole-2-carboxylic acid, 98%
C9H7NO2