EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
A characterisation study on the application of inverted lyotropic phases for subcutaneous drug release.
PMID 20026201
Abstract
An experimental characterisation of lipid mixtures consisting of inverted hexagonal and inverted cubic phases composed of soybean phosphatidylcholine (SPC) and glycerol dioleate (GDO) was performed. The release of five chromophores of varying lipophilicity, used as model drugs, was investigated. Two experimental setups were applied: one based on maintaining sink condition, while a constant volume release medium was employed for the other. For neither setup, no correlation between the model drug lipophilicity and the polarity of the carrier matrix was found. However, the lipid phases showed a prolonged release, spanning weeks, of the model drugs, which exhibit lipophilicity values ranging by four orders of magnitude.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D8894
Dioleoylglycerol, ≥99%, mixture of 1,3- and 1,2-isomers, liquid
C39H72O5