EMAIL THIS PAGE TO A FRIEND
Alcoholism, clinical and experimental research
Differential effects of ethanol on serum GABAergic 3alpha,5alpha/3alpha,5beta neuroactive steroids in mice, rats, cynomolgus monkeys, and humans.
PMID 20028362
Abstract
Acute ethanol administration increases plasma and brain levels of progesterone and deoxycorticosterone-derived neuroactive steroids (3alpha,5alpha)-3-hydroxypregnan-20-one (3alpha,5alpha-THP) and (3alpha,5alpha)-3,21-dihydroxypregnan-20-one (3alpha,5alpha-THDOC) in rats. However, little is known about ethanol effects on GABAergic neuroactive steroids in mice, nonhuman primates, or humans. We investigated the effects of ethanol on plasma levels of 3alpha,5alpha- and 3alpha,5beta-reduced GABAergic neuroactive steroids derived from progesterone, deoxycorticosterone, dehydroepiandrosterone, and testosterone using gas chromatography-mass spectrometry. Serum levels of GABAergic neuroactive steroids and pregnenolone were measured in male rats, C57BL/6J and DBA/2J mice, cynomolgus monkeys, and humans following ethanol administration. Rats and mice were injected with ethanol (0.8 to 2.0 g/kg), cynomolgus monkeys received ethanol (1.5 g/kg) intragastrically, and healthy men consumed a beverage containing 0.8 g/kg ethanol. Steroids were measured after 60 minutes in all species and also after 120 minutes in monkeys and humans. Ethanol administration to rats increased levels of 3alpha,5alpha-THP, 3alpha,5alpha-THDOC, and pregnenolone at the doses of 1.5 g/kg (+228, +134, and +860%, respectively, p < 0.001) and 2.0 g/kg (+399, +174, and +1125%, respectively, p < 0.001), but not at the dose of 0.8 g/kg. Ethanol did not alter levels of the other neuroactive steroids. In contrast, C57BL/6J mice exhibited a 27% decrease in serum 3alpha,5alpha-THP levels (p < 0.01), while DBA/2J mice showed no significant effect of ethanol, although both mouse strains exhibited substantial increases in precursor steroids. Ethanol did not alter any of the neuroactive steroids in cynomolgus monkeys at doses comparable to those studied in rats. Finally, no effect of ethanol (0.8 g/kg) was observed in men. These studies show clear species differences among rats, mice, and cynomolgus monkeys in the effects of ethanol administration on circulating neuroactive steroids. Rats are unique in their pronounced elevation of GABAergic neuroactive steroids, while this effect was not observed in mice or cynomolgus monkeys at comparable ethanol doses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2016
3α,21-Dihydroxy-5α-pregnan-20-one, ≥95%
C21H34O3