EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and application of a photoaffinity analog of dehydroepiandrosterone (DHEA).
PMID 20031404
Abstract
We have synthesized an analog of dehydroepiandrosterone (DHEA, 1) containing both a benzophenone (BP) and a biotin (Bt) group (DHEA-BP-Bt, 8). Compound 8 was prepared by functionalization on C-17 of 1. Biocytin was reacted with 4-benzoylbenzoic acid and the product was condensed with 1 containing a diamine-hexane linker. We detected specific protein bands of approximately 55, 80, and 150 kDa by SDS-PAGE analysis of vascular endothelial cell plasma membranes which had been photoirradiated in the presence of 8.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
775924
4-Benzoylbenzoic acid N-succinimidyl ester, 97%
C18H13NO5