EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Regioselective synthesis of new 2-(E)-cyano(thiazolidin-2-ylidene)thiazoles.
PMID 20032863
Abstract
Cyclocondensation of 2-[bis(methylthio)methylene]malononitrile (1) and cysteamine (2) afforded 2-(thiazolidin-2-ylidene)malononitrile (3). This compound on treatment with NaSH gave the corresponding thioamide derivative 4a in a regioselective manner on the basis of its single crystal X-ray diffraction analysis. Reaction of this compound with several alpha-bromocarbonyl compounds gave new 2-(E)-cyano(thiazolidin-2-ylidene)thiazoles 5a-g.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
161527
Sodium hydrosulfide hydrate
HNaS · xH2O