EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and antioxidant properties of enone core based dendrimers with carbazole as surface group.
PMID 20034709
Abstract
Synthesis of enone core based dendrimers with carbazole as surface group has been achieved. All the synthesized dendrimers showed excellent antioxidant behavior with commercially available 1,1-diphenyl-2-picryl hydrazyl (DPPH).