EMAIL THIS PAGE TO A FRIEND
Organic letters
Synthesis of tetrahydropyrroloiminoquinone alkaloids based on electrochemically generated hypervalent iodine oxidative cyclization.
PMID 20039698
Abstract
An approach to the synthesis of the tetrahydropyrroloiminoquinone alkaloids has been developed and applied to the preparation of N-1-beta-D-ribofuranosyltetrahydropyrroloiminoquinones. The strategy utilizes oxidative cyclization of aryl-methoxyamides by hypervalent iodine to construct the quinoline framework shared by members of this alkaloid family. The hypervalent iodine oxidant is generated in situ by anodic oxidation of iodobenzene.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I7632
Iodobenzene, 98%
C6H5I