EMAIL THIS PAGE TO A FRIEND
International journal of clinical pharmacology and therapeutics
Protracted deep coma after bromazepam poisoning.
PMID 20040343
Abstract
Bromazepam intoxication is very common but surprisingly rarely reported. We describe the case of a 73-year-old woman who suffered from a prolonged coma after acute self poisoning with bromazepam (serum concentration of 2,000 ng/ml at admission, 2 - 10 hours after ingestion of up to 180 mg) and zolpidem (900 ng/ml at admission). Only the former lasted at toxic concentrations. Recovery of consciousness allowed extubation on Day 16. Repeat-dose activated charcoal (25 g every 6 h from Day 14 to 16) resulted in minimal effects on bromazepam grossly estimated kinetics. Despite its relatively low theoretic half-life, bromazepam may induce a prolonged life-threatening coma, even in the absence of renal or hepatic failure.