EMAIL THIS PAGE TO A FRIEND
Anesthesia and analgesia
The effects of the phosphodiesterase inhibitor olprinone on global cerebral ischemia.
PMID 20042441
Abstract
The phosphodiesterase III inhibitor olprinone has been confirmed to improve myocardial function and increase cerebral blood flow; therefore, if olprinone exerts direct neuroprotective effects against global cerebral ischemia to the same degree as cilostazol, olprinone could be useful for cerebral resuscitation after cardiac arrest. We examined whether olprinone directly protected neuronal cells from global cerebral ischemia both in vivo and in vitro. In a rat model of 10-minute global cerebral ischemia induced by 4-vessel occlusion, 0.3, 3, or 30 microg x kg(-1) x min(-1) olprinone or saline was infused for a periischemic period of 40 minutes (n = 6 for each group). Hippocampal CA1 neuronal cells were then counted 3 days after reperfusion, and the phosphorylation of cyclic adenosine 3'5'-monophosphate response element-binding protein was examined using Western blotting analyses of specimens obtained 15 minutes after reperfusion. In vitro, cultured cerebral neurons were exposed to 4 hours of hypoxia and glucose deprivation and then 24 hours of recovery in the absence or presence of olprinone (10(-11)-10(-5) mol x L(-1)). Cell viability was measured using the Cell Counting Kit-8 (Dojindo Molecular Technologies, Gaithersburg, MD). In the rat model of global ischemia, the number of surviving CA1 neurons counted under a microscopic field in the 30 microg x kg(-1) x min(-1) olprinone-treated group (49.9 +/- 9.2) was significantly higher than that in the saline infusion control group (7.2 +/- 3.4), and olprinone treatment increased the phosphorylation of cyclic adenosine 3'5'-monophosphate response element-binding protein. The survival fraction of the neuronal cells cultured in the presence of olprinone was also significantly higher than that of cells cultured in the absence of olprinone in a dose-dependent manner. Our study successfully demonstrated, for the first time, that olprinone had a protective effect on neuronal cells in vitro and in vivo, especially against global cerebral ischemia. These results suggest that olprinone might be useful for the treatment of patients experiencing global cerebral ischemia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O7389
Olprinone hydrochloride, ≥98% (HPLC), solid
C14H10N4O · HCl