EMAIL THIS PAGE TO A FRIEND
American journal of health-system pharmacy : AJHP : official journal of the American Society of Health-System Pharmacists
Effect of an educational initiative on physicians' use of weight-based fosphenytoin loading doses in the emergency department.
PMID 20044359
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0117
Fosphenytoin disodium salt hydrate, ≥98% (HPLC)
C16H13N2Na2O6P · xH2O