EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Proteasome inhibition in human breast cancer cells with high catechol-O-methyltransferase activity by green tea polyphenol EGCG analogs.
PMID 20045338
Abstract
A pro-drug 8 of a synthetic analog 7 is more active in its antiproliferative activity against human breast cancer MDA-MB-231 cells possessing high catechol-O-methyltransferase (COMT) activity than the pro-drugs of EGCG and the analog 5. The higher activity of 8 is attributed to it not being a substrate of COMT.
Related Materials