EMAIL THIS PAGE TO A FRIEND
EMBO molecular medicine
Methotrexate induces oxidative DNA damage and is selectively lethal to tumour cells with defects in the DNA mismatch repair gene MSH2.
PMID 20049736
Abstract
Mutations in the MSH2 gene predispose to a number of tumourigenic conditions, including hereditary non-polyposis colon cancer (HNPCC). MSH2 encodes a protein in the mismatch repair (MMR) pathway which is involved in the removal of mispairs originating during replication or from damaged DNA. To identify new therapeutic strategies for the treatment of cancer arising from MMR deficiency, we screened a small molecule library encompassing previously utilized drugs and drug-like molecules to identify agents selectively lethal to cells lacking functional MSH2. This approach identified the drug methotrexate as being highly selective for cells with MSH2 deficiency. Methotrexate treatment caused the accumulation of potentially lethal 8-hydroxy-2'-deoxyguanosine (8-OHdG) oxidative DNA lesions in both MSH2 deficient and proficient cells. In MSH2 proficient cells, these lesions were rapidly cleared, while in MSH2 deficient cells 8-OHdG lesions persisted, potentially explaining the selectivity of methotrexate. Short interfering (si)RNA mediated silencing of the target of methotrexate, dihydrofolate reductase (DHFR), was also selective for MSH2 deficiency and also caused an accumulation of 8-OHdG. This suggested that the ability of methotrexate to modulate folate synthesis via inhibition of DHFR, may explain MSH2 selectivity. Consistent with this hypothesis, addition of folic acid to culture media substantially rescued the lethal phenotype caused by methotrexate. While methotrexate has been used for many years as a cancer therapy, our observations suggest that this drug may have particular utility for the treatment of a subset of patients with tumours characterized by MSH2 mutations.