EMAIL THIS PAGE TO A FRIEND
Journal of biomedical nanotechnology
Improving oral absorption of Salmon calcitonin by trimyristin lipid nanoparticles.
PMID 20055109
Abstract
Solid lipid nanoparticles (SLN) composed of trimyristin (solid lipid) and poloxamer 407 (surfactant) were prepared by a w/o/w emulsion technique for the incorporation of Salmon calcitonin, and further explored as protein carriers for oral delivery. Trimyristin SLN showed a mean size diameter of 200 nm with an association efficiency for calcitonin of approx. 86%. The morphology of SLN was investigated by cryo-SEM and by AFM, revealing spheroid shape SLN with a smooth surface. The in vitro release of calcitonin occurred for a period of 8 h, under both gastric and intestinal simulated pH conditions, predicting suitable properties for oral administration. The pharmacological activity of the protein was evaluated following oral dosage of calcitonin-loaded SLN in rats. SLN lowered the basal blood calcium levels by up to 20% with 500 IU/kg dose sustaining hypocalcaemia over 8 h. The results indicate that incorporation of Salmon calcitonin into trimyristin SLN is a key factor for the improvement of the efficiency of such carriers for oral delivery of proteins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T5141
Glyceryl trimyristate, ≥99%
C45H86O6