EMAIL THIS PAGE TO A FRIEND
Yao xue xue bao = Acta pharmaceutica Sinica
[A new trincallane derivative from Salacia hainanensis Chun et How].
PMID 20055135
Abstract
Chemical constituents of the roots and stem of Salacia hainanensis Chun et How were isolated and purified with column chromatography on silica gel, Sephadex LH-20 and preparative HPLC. Their structures were elucidated based on physicochemical and spectral spectroscopic analysis. Depending on the activities of anti-alpha-glucosidase and inhibiting AGEs (advanced glycation end products, AGEs) formation in vitro, nine compounds were identified as 26, 27-dihydroxy-7, 24-tirucalladien-3-one (1), abruslactone A (2), lupeol (3), 21alpha, 30-dihydroxy-D: A-friedooleanan-3-one (4), 15alpha-hydroxyfriedelan-3-one (5), friedelin (6), mangiferin (7), epicatechin (8) and beta-sitosterol (9), separately. Among them, compound 1 is a new compound, and compound 2 was isolated from the Salacia genus for the first time, while, compounds 3, 4, 5, 8 were obtained from this plant for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
855022
Friedelin, technical grade
C30H50O