EMAIL THIS PAGE TO A FRIEND
Inorganic chemistry
Titanocene difluorides with improved cytotoxic activity.
PMID 20055428
Abstract
Titanocene difluorides can be obtained by halide metathesis of the respective titanocene dichlorides with trimethyltin fluoride (Me(3)SnF), giving access to a new class of cytotoxic active substances. Furthermore, an improved method for the synthesis of diaryl-substituted titanocene dichlorides is presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
234826
Bis(cyclopentadienyl)titanium(IV) dichloride, 97%
C10H10Cl2Ti