EMAIL THIS PAGE TO A FRIEND
Acta paediatrica (Oslo, Norway : 1992)
Outcome of very preterm infants with Mycoplasma/Ureaplasma airway colonization treated with josamycine.
PMID 20055775
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
59983
Josamycin, ≥90% (HPLC)
C42H69NO15
Y0000041
Josamycin, European Pharmacopoeia (EP) Reference Standard
C42H69NO15