Clinical journal of the American Society of Nephrology : CJASN
A randomized trial of cholecalciferol versus doxercalciferol for lowering parathyroid hormone in chronic kidney disease.
PMID 20056760
The optimal treatment of secondary hyperparathyroidism in chronic kidney disease (CKD) is unknown. We conducted a randomized, blinded, 3-month trial in vitamin D-deficient CKD stage 3 and 4 patients with parathyroid hormone (PTH) values above the Kidney Disease Outcomes Quality Initiative target, comparing cholecalciferol (4000 IU/d x 1 month, then 2000 IU/d; n = 22) to doxercalciferol (1 microg/d; n = 25). There was no difference in baseline demographics or lab tests, except a slightly higher estimated GFR (eGFR) in the doxercalciferol group. There was a significant increase in vitamin D level in the cholecalciferol group (14 +/- 6 to 37 +/- 10 ng/ml; P < 0.001) but no change in the doxercalciferol group. The PTH decreased by 27% +/- 34% in the doxercalciferol group (P = 0.002) and decreased by 10% +/- 31% in the cholecalciferol group (P = 0.16), but the difference between treatments was NS (P = 0.11). Similar results were found when absolute PTH change from baseline to end point was analyzed in a repeated-measures ANOVA model. The serum calcium and urine calcium excretions were not different. Additional non-mineral-related end points, albuminuria, and BP were evaluated, and although trends were present, this did not reach significance. This prospective, randomized trial demonstrated a within-group reduction in PTH in the doxercalciferol-treated patients but no significant difference between the doxercalciferol and cholecalciferol patients. Larger, long-term studies are needed to demonstrate efficacy of mineral-related and non-mineral-related end points and safety.
Related Materials
Product #
Molecular Formula
Add to Cart
Doxercalciferol, ≥98% (HPLC), solubility: >10 mg/mL in DMSO