EMAIL THIS PAGE TO A FRIEND
AAPS PharmSciTech
In vitro and in vivo evaluation of proniosomes containing celecoxib for oral administration.
PMID 20058106
Abstract
The objectives of this research were to prepare celecoxib proniosomes and evaluate the influence of proniosomal formulation on the oral bioavailability of the drug in human volunteers. A new proniosomal delivery system for a poorly water-soluble drug such as celecoxib was developed and subjected to in vitro and in vivo studies. Proniosomes were prepared by sequential spraying method, which consisted of cholesterol, span 60, and dicetyl phosphate in a molar ratio of 1:1: 0.1, respectively. The average entrapment percent of celecoxib proniosome-derived niosomes was about 95%. The prepared proniosomes showed marked enhancement in the dissolution of celecoxib as compared to pure drug powder. The bioavailability of 200 mg single dose of both celecoxib proniosomal formulation and a conventional marketed celecoxib capsule was studied in human volunteers. The obtained results show that the proniosomal formulation significantly improved the extent of celecoxib absorption than conventional capsule. The mean relative bioavailability of the proniosomal formulation to the conventional capsule was 172.06 +/- 0.14%. The mean T (max) for celecoxib was prolonged when given as proniosomal capsule. There was no significant difference between the values of K (el) and t (1/2) for both celecoxib preparations. In conclusion, the proniosomal oral delivery system of celecoxib with improved bioavailability was established.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S7010
Span® 60