EMAIL THIS PAGE TO A FRIEND
Experimental Cell Research
Up- or downregulation of tescalcin in HL-60 cells is associated with their differentiation to either granulocytic or macrophage-like lineage.
PMID 20060826